Läs mer
Fenhexamid, TRC
Produktkod. 30408427
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30408427 50 mg
1 options
Produktkod. 30408427 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF24765050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fenhexamid
CAS 126833-17-8
Molekylformel C14 H17 Cl2 N O2
Kvantitet 50 mg
Synonym N-(2,3-Dichloro-4-hydroxyphenyl)-1-methylcyclohexanecarboxamide, Decree, Elevate, Fenhexamid, KBR 2738, Teldor, Cyclohexanecarboxamide, N-(2,3-dichloro-4-hydroxyphenyl)-1-methyl-
InChI formel InChI=1S/C14H17Cl2NO2/c1-14(7-3-2-4-8-14)13(19)17-9-5-6-10(18)12(16)11(9)15/h5-6,18H,2-4,7-8H2,1H3,(H,17,19)
LEDER CC1(CCCCC1)C(=O)Nc2ccc(O)c(Cl)c2Cl
IUPAC-namn N-(2,3-dichloro-4-hydroxyphenyl)-1-methylcyclohexane-1-carboxamide
Molekylvikt (g/mol) 302.2
Formel vikt 301.0636