Läs mer
Fenoldopam Hydrobromide, TRC
Produktkod. 30405346
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30405346 10 mg
1 options
Produktkod. 30405346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF24879010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fenoldopam Hydrobromide
CAS 67287-54-1
Molekylformel C16 H16 Cl N O3 . H Br
Kvantitet 10 mg
Synonym 6-Chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-1H-3-benzazepine-7,8-diol Hydrobromide, (±)-Fenoldopam Hydrobromide, (±)-SKF 82526 Hydrobromide, SKF 82526 Hydrobromide
InChI formel InChI=1S/C16H16ClNO3.BrH/c17-15-11-5-6-18-8-13(9-1-3-10(19)4-2-9)12(11)7-14(20)16(15)21;/h1-4,7,13,18-21H,5-6,8H2;1H/t13-;/m1./s1
LEDER Br.Oc1ccc(cc1)[C@H]2CNCCc3c(Cl)c(O)c(O)cc23
IUPAC-namn (1R)-6-chloro-1-(4-hydroxyphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;hydrobromide
Molekylvikt (g/mol) 386.67
Formel vikt 385.008