Läs mer
Fenoterol Hydrobromide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Fenoterol Hydrobromide |
| CAS | 1944-12-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C17 H21 N O4 . Br H |
| Kvantitet | 50 mg |
| Synonym | Fenoterol hydrobromide, (1RS)-1-(3,5-Dihydroxyphenyl)-2-[[(1RS)-2-(4-hydroxyphenyl)-1-methylethyl]amino]ethanol hydrobromide, 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide (1:1), 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide (9CI), Benzyl alcohol, 3,5-dihydroxy-α-[[(p-hydroxy-α-methylphenethyl)amino]methyl]-, hydrobromide (8CI), 1-(3,5-Dihydroxyphenyl)-2-[2-(3-p-hydroxyphenyl)propylamino]ethanol hydrobromide, 1-(p-Hydroxyphenyl)-2-(β-3',5'-dihydroxyphenyl-β-hydroxy)ethylaminopropane hydrobromide, 3,5-Dihydroxy-α-[(p-hydroxy-α-methylphenethyl)amino]methylbenzyl alcohol hydrobromide, 3,5-Dihydroxy-α-[[(p-hydroxy-α-methylphenethyl)amino]methyl]benzyl alc. monohydrobromide, Airum, Berotec, Dosberotec, Fenoterol bromide, Fenoterol hydrobromide, Partusisten, Phenoterol hydrobromide, Th 1165a, 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide, (R*,R*)-(±)-, 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide, (R*,R*)- |
| InChI formel | InChI=1S/C17H21NO4.BrH/c1-11(6-12-2-4-14(19)5-3-12)18-10-17(22)13-7-15(20)9-16(21)8-13;/h2-5,7-9,11,17-22H,6,10H2,1H3;1H/t11-,17+;/m1./s1 |
| LEDER | Br.C[C@H](Cc1ccc(O)cc1)NC[C@H](O)c2cc(O)cc(O)c2 |
| IUPAC-namn | 5-[(1R)-1-hydroxy-2-[[(2R)-1-(4-hydroxyphenyl)propan-2-yl]amino]ethyl]benzene-1,3-diol;hydrobromide |
| Molekylvikt (g/mol) | 384.26 |
| Visa mer |