Läs mer
Fenothiocarb (~90%), TRC
Produktkod. 30406540
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30406540 10 mg
1 options
Produktkod. 30406540 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF24872010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fenothiocarb
CAS 62850-32-2
Molekylformel C13 H19 N O2 S
Kvantitet 10 mg
Synonym ENT 27,579, 2-(Ethylthio)phenyl methylcarbamate
InChI formel InChI=1S/C13H19NO2S/c1-14(2)13(15)17-11-7-6-10-16-12-8-4-3-5-9-12/h3-5,8-9H,6-7,10-11H2,1-2H3
LEDER CN(C)C(=O)SCCCCOc1ccccc1
IUPAC-namn S-(4-phenoxybutyl) N,N-dimethylcarbamothioate
Molekylvikt (g/mol) 253.36
Formel vikt 253.1136