Läs mer
Fenpiverinium Bromide, TRC
Produktkod. 30009308
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009308 100 mg
30008455 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009308 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF249000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 125-60-0
Molekylformel C22 H29 N2 O . Br
Molekylvikt (g/mol) 417.38
Synonym Piperidinium, 1-(4-amino-4-oxo-3,3-diphenylbutyl)-1-methyl-, bromide (1:1), 1-(3-Carbamoyl-3,3-diphenylpropyl)-1-methylpiperidinium bromide (6CI,7CI), Piperidinium, 1-(3-carbamoyl-3,3-diphenylpropyl)-1-methyl-, bromide (8CI), Piperidinium, 1-(4-amino-4-oxo-3,3-diphenylbutyl)-1-methyl-, bromide (9CI), 12494Hoechst, 2,2-Diphenyl-2-(2-piperidinoethyl)acetamide methyl bromide, 2-(Diphenylpiperidinoethyl)acetamide methobromide, 2-(Diphenylpiperidinoethyl)acetamide methyl bromide, Baralgin-Amid, Baralgin-Amide, Bulgar amide, Fenpipramide methobromide, Fenpiverinium bromide, Hoechst 12494, Resantin, U 0385, α,α-Diphenyl-1-piperidinebutyramide methobromide, α,α-Diphenyl-γ-piperidylbutyramide methobromide
IUPAC-namn 4-(1-methylpiperidin-1-ium-1-yl)-2,2-diphenylbutanamide;bromide
LEDER [Br-].C[N+]1(CCC(C(=O)N)(c2ccccc2)c3ccccc3)CCCCC1
Specifikationer
Kemiskt namn eller material Fenpiverinium Bromide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C22H28N2O.BrH/c1-24(16-9-4-10-17-24)18-15-22(21(23)25,19-11-5-2-6-12-19)20-13-7-3-8-14-20;/h2-3,5-8,11-14H,4,9-10,15-18H2,1H3,(H-,23,25);1H
Formel vikt 416.1463
Procent renhet >95%