Läs mer
Fenspiride hydrochloride, MedChemExpress
Produktkod. 16467996
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16467996 100 mg
1 options
Produktkod. 16467996 Leverantör Medchem Express Leverantörsnummer HYA0027
MedChemExpress Fenspiride, an orally active non-steroidal antiinflammatory agent, is an antagonist of H1-histamine receptor. Fenspiride inhibites phosphodiesterase 3 (PDE3), phosphodiesterase 4 (PDE4) and phosphodiesterase 5 (PDE5) activities with -log IC50 values of 3.44, 4.16 and approximately 3.8, respectively. Fenspiride can be used for the research of respiratory diseases.
Fenspiride, an orally active non-steroidal antiinflammatory agent, is an antagonist of H1-histamine receptor. Fenspiride inhibites phosphodiesterase 3 (PDE3), phosphodiesterase 4 (PDE4) and phosphodiesterase 5 (PDE5) activities with -log IC50 values of 3.44, 4.16 and approximately 3.8, respectively. Fenspiride can be used for the research of respiratory diseases.
Specifikationer
Kemiskt namn eller material Fenspiride hydrochloride
Fysisk form Solid
CAS 8-7-5053
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C15H21ClN2O2
För användning med (applikation) COVID-19-immunoregulation
Löslighetsinformation H2O : 50 mg/mL (168.47 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (112.30 mM; Need ultrasonic)
LEDER O=C1OC2(CCN(CCC3=CC=CC=C3)CC2)CN1.[H]Cl
Molekylvikt (g/mol) 296.79
Formel vikt 296.79
Procent renhet 99.11%