Läs mer
Fenthoxon Sulfoxide, TRC
Produktkod. 30407594
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30407594 25 mg
1 options
Produktkod. 30407594 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF27901525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fenthoxon Sulfoxide
CAS 6552-13-2
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H15 O5 P S
Kvantitet 25 mg
Synonym Fenthion Oxon Sulfoxide, Phosphoric acid, dimethyl 3-methyl-4-(methylsulfinyl)phenyl ester, Phosphoric acid, dimethyl 4-(methylsulfinyl)-m-tolyl ester (7CI,8CI), Fenoxon sulfoxide, Fenthion oxon sulfoxide, Fenthion-oxon-sulfoxide
InChI formel InChI=1S/C10H15O5PS/c1-8-7-9(5-6-10(8)17(4)12)15-16(11,13-2)14-3/h5-7H,1-4H3
LEDER COP(=O)(OC)Oc1ccc(c(C)c1)S(=O)C
IUPAC-namn dimethyl (3-methyl-4-methylsulfinylphenyl) phosphate
Molekylvikt (g/mol) 278.2619
Formel vikt 278.0378