missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ FER Polyclonal Antibody
GREENER_CHOICE

Produktkod. 15994815
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15994815 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15994815 Leverantör Invitrogen™ Leverantörsnummer PA579248

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, Rat Testis Tissue, Rat Ovary Tissue. IHC: Human Intestinal Cancer Tissue, Rat Intestine Tissue.

FER is a member of the FPS/FES family of non-receptor tyrosine kinases. It regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors. FER is found in the cytoplasm and in the nucleus, where it was associated with the chromatin fraction. Its ubiquitous expression and its subcellular location indicate that it may be involved in key regulatory processes. The group IV cytoplasmic protein-tyrosine kinase Fer has been linked to cellular signaling responses to many different stimuli, including growth factors and cytokines. Data suggest a biological role for Fer in regulation of leukocyte recruitment during the innate immune response.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen FER
Användningsområden Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Koncentration 500 μg/mL
Konjugera Unconjugated
Formulering PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gen FER
Genaccessionsnr. P09760, P16591
Gene Alias AV082135; C330004K01Rik; c-FER; EC 2.7.10.2; Feline encephalitis virus-related kinase FER; FER; fer (fms/fps related) protein kinase; fer (fms/fps related) protein kinase, testis specific 2; fer (fps/fes related) tyrosine kinase; FER tyrosine kinase; Fert; Fert2; Flk; Flk_retired; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; p47; p94-Fer; phosphoprotein NCP94; PPP1R74; protein phosphatase 1, regulatory subunit 74; proto-oncogene c-Fer; proto-oncogene tyrosine-protein kinase Fer; TYK3; Tyrosine kinase 3; tyrosine protein kinase FLK; tyrosine-protein kinase Fer; Tyrosine-protein kinase FLK
Gensymboler FER
Värd art Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human FER (521-536aa FSNIPQLIDHHYTTKQ).
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 2241, 301737
Målarter Human, Rat
Innehåll och lagring -20°C
Produkttyp Antibody
Form Lyophilized
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.