missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FER Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25287-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Protein kinases constitute a large superfamily of enzymes with key regulatory functions in nearly all signal transmission processes of eukaryotic cells. FER is found in the cytoplasm and in the nucleus, where it was associated with the chromatin fraction. Its ubiquitous expression and its subcellular location indicate that it may be involved in key regulatory processes. The group IV cytoplasmic protein-tyrosine kinase Fer has been linked to cellular signaling responses to many different stimuli, including growth factors and cytokines. Data suggest a biological role for Fer in regulation of leukocyte recruitment during the innate immune response.Specifications
FER | |
Polyclonal | |
Unconjugated | |
FER | |
FER, p94 Fer, Proto oncogene c Fer, TYK3, Tyrosine kinase 3, Tyrosine protein kinase Fer | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14158, 2241 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16591, P70451 | |
FER | |
FER Fusion Protein Ag18173 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |