Learn More
Thermo Scientific™ dNTP Mix (25 mM each)
Thermo Scientific™ dNTP Mix, 25mM each of dATP, dTTP, dCTP and dGTP.
Brand: Thermo Scientific™ R1122
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ dNTP Mix contains 25mM each of dATP, dTTP, dCTP and dGTP.
For direct use in PCR, reverse transcription and other applications.Specifications
Liquid | |
>99% | |
Standard PCR |
Unlabeled | |
5 x 1 mL | |
25 mM |
For Research Use Only. Not for use in diagnostic procedures.