Learn More
Thermo Scientific™ Lambda DNA/EcoRI Marker
Perform sizing and approximate quantification of large double-stranded DNA fragments in agarose gels.
Brand:  Thermo Scientific™ SM0281
Additional Details : Weight : 1.51950kg
Product Code. 11803963
  • 590.00 SEK / Pack of 5
In Stock
to see stock.
Add to basket
Description
Description
Lambda DNA is digested to completion with the appropriate Thermo Scientitific restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively, it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction (see Recources for Protocols).
Highlights
  • Sizing and approximate quantification of large DNA fragments
  • Sharp bands
  • Supplied with loading dye for sample DNA
Specifications
Specifications
6X
Xylene Cyanol FF, Bromophenol Blue
No
Sustainable packaging
Agarose
DNA/ECORI Marker
Agarose Gels