Learn More
Thermo Scientific™ pBR322 DNA/BsuRI (HaeIII) Marker
Perform sizing and approximate quantification of small double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels.
Brand: Thermo Scientific™ SM0271
374.68 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00900kg
Description
pBR322 DNA/BsuRI (HaeIII) Marker is recommended for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels. pBR322 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends depending on the restriction enzyme used for the marker's preparation
The marker can be labeled radioactively with T4 Polynucleotide Kinase.
Highlights
- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
Specifications
Agarose | |
No | |
Sustainable packaging |
DNA/BSURI (HAEIII) Marker | |
Agarose Gels |
For Research Use Only. Not for use in diagnostic procedures.