Läs mer
Ferroprotoporphyrin IX, TRC
Produktkod. 30133896
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133896 50 mg
30137591 25 mg
30005205 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133896 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF30761050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14875-96-8
Molekylformel C34H32FeN4O42
Molekylvikt (g/mol) 616.49
Synonym 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic Acid Ferrous Complex, Ferroheme, Hem Fe, Heme, Heme B, Hydrogen [7,12-Diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropanoato(4-)-κN21,κN22,κN23,κN24]ferrate (heme), Iron Protoporphyrin, Iron Protoporphyrin IX, Iron(II) Protoporphyrin IX, NSC 267100, Protoheme, Protoheme IX, Reduced Hematin, [Dihydrogen 3,7,12,17-tetramethyl-8,13-divinyl-2,18-porphinedipropionato(2-)]iron, (SP-4-2)-[7,12-diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropanoato(4-)-N21,N22,N23,N24]-Ferrate(2-) Dihydrogen, Protoporphyrin Iron Complex, 7,12-diethenyl-3,8,13,17-tetramethyl-21H,23H-Porphine-2,18-dipropanoic Acid Iron Complex, (SP-4-2)-[7,12-diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropanoato(4-)-κN21,κN22,κN23,κN24]-ferrate(2-) Dihydrogen;
LEDER OC(CCC1=C(C=C2[N]3=C4C(C)=C2CCC(O)=O)[N]([Fe]35[N]6=C7C=C(C(C=C)=C8C)[N]5C8=C4)C(C=C6C(C=C)=C7C)=C1C)=O
Specifikationer
Kemiskt namn eller material Ferroprotoporphyrin IX
Kvantitet 50 mg
InChI formel InChI=1S/C34H34N4O4.Fe/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);/q;+2/p-2/b25-13?,26-13-,27-14?,28-15-,29-14+,30-15?,31-16-,32-16?;
Formel vikt 616.1773