Läs mer
Ferrous Sulphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ferrous sulfate |
| CAS | 7720-78-7 |
| Molekylformel | Fe . O4 S |
| Kvantitet | 25 g |
| Synonym | Sulfuric acid, iron(2+) salt (1:1), Combiron, Copperas, Duretter, Duroferon, Exsiccated ferrous sulfate, Exsiccated ferrous sulphate, Feofol Spansule, Feosol, Feospan, Fer-In-Sol, Ferlim, Fero-Folic 500, Ferogradumet, Ferralyn, Ferro-Gradumet, Ferro-Thero |
| InChI formel | InChI=1S/Fe.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2 |
| LEDER | [Fe+2].[O-]S(=O)(=O)[O-] |
| IUPAC-namn | iron(2+);sulfate |
| Molekylvikt (g/mol) | 151.908 |
| Formel vikt | 151.887 |