Learn More
Invitrogen™ FES (FPS) Recombinant Human Protein
FES (FPS) is a tyrosine protein kinase that is required for maintenance of cellular transformation.
Brand: Invitrogen™ PR5068B
Additional Details : Weight : 0.01000kg
Description
The chromosomal location of FES (FPS) is linked to a specific translocation event identified in patients with acute promyelocytic leukemia, and it is also involved in normal hematopoiesis.
Specifications
NP_001996 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 4mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
FES (FPS) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
98.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
FER Kinase Family | |
Recombinant | |
Baculovirus |