missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fetuin, From Fetal Bovine Serum

Fetuin is a protein containing 12 Cys residues, all involved in disulfide bridges.

Brand:  MP Biomedicals™ 02104874.1

Additional Details : CAS Number : 9014-81-7 Weight : 0.01000kg

 View more versions of this product

Product Code. 11431752

  • 2586.40 SEK / 100mg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Formulation: Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%

Applications:

Fetuin bovine is a Protein with the ability to bind to the insulin receptor and inhibit insulin action, Fetuin has been used in studies of type 2 diabetes mellitus (T2DM), In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients.

Specifications

Specifications

Type 2 Diabetes Mellitus (T2DM) Studies, Cell Attachment and Proliferation
Fetuin Protein
100mg
Powder
Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%
Ammonium Sulfate Fractionation
Fetal Bovine Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers