Learn More
MP Biomedicals™ Fetuin, From Fetal Bovine Serum
Fetuin is a protein containing 12 Cys residues, all involved in disulfide bridges.
Brand: MP Biomedicals™ 02104874.2
Additional Details : CAS Number : 9014-81-7 Weight : 0.01000kg
Description
- Formulation: Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%
Applications:
Fetuin bovine is a Protein with the ability to bind to the insulin receptor and inhibit insulin action, Fetuin has been used in studies of type 2 diabetes mellitus (T2DM), In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients.
Specifications
Fetuin bovine is a protein with the ability to bind to the insulin receptor and inhibit insulin action, Fetuin has been used in studies of type 2 diabetes mellitus (T2DM), In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients, Fetuin has also been used in studies to demonstrate its protective role in the development of arterial calcification. Fetuin also contributes to the attachment and spreading of cells in culture medium | |
Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7% | |
Ammonium Sulfate Fractionation | |
Fetal Bovine Serum |
Powder | |
Fetuin Protein | |
250mg |