missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEV Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566944
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MRQSGASQPLL INMYLPDPVG DGLFKDGKNP SWGPLSPAVQ KGSGQ Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
This gene belongs to the ETS transcription factor family. ETS family members have a highly conserved 85-amino acid ETS domain that binds purine-rich DNA sequences. The alanine-rich C-terminus of this gene indicates that it may act as a transcription repressor. This gene is exclusively expressed in neurons of the central serotonin (5-HT) system, a system implicated in the pathogeny of such psychiatric diseases as depression, anxiety, and eating disorders. In some types of Ewing tumors, this gene is fused to the Ewing sarcoma (EWS) gene following chromosome translocations. [provided by RefSeq, Jul 2008].Specifications
FEV | |
Polyclonal | |
Unconjugated | |
FEV | |
FEV (ETS oncogene family); HSRNAFEV; PET-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54738 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99581 | |
FEV | |
Recombit Human FEV | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only