Läs mer
Fexofenadine EP Impurity C, Mikromol™
Produktkod. 30049063
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049063 100 mg
30050286 25 mg
30460093 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30049063 Leverantör Mikromol Leverantörsnummer MM0507.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Fexofenadine EP Impurity C
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1RS)-4-(4-(Hydroxydiphenylmethyl)piperidin-1-yl)-1-(4-(1-methylethyl)phenyl)butan-1-ol
CAS 185066-37-9
Analyt- eller komponentnamn (1RS)-4-(4-(Hydroxydiphenylmethyl)piperidin-1-yl)-1-(4-(1-methylethyl)phenyl)butan-1-ol
Frakt skick Room Temperature
Molekylformel C31 H39 N O2
Certifikat och efterlevnad ISO 9001
Synonym (1RS)-4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]-1-[4-(1-methylethyl)phenyl]butan-1-ol, Fexofenadine Hydrochloride Imp. C (EP), Fexofenadine Imp. C (EP), Fexofenadine USP Related Compound C, Fexofenadine USP RC C, (1RS)-4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]-1-[4-(1-methylethyl)phenyl]butan-1-ol, 1-Piperidinebutanol, 4-(hydroxydiphenylmethyl)-alpha-[4-(1-methylethyl)phenyl]-, 4-(Hydroxydiphenylmethyl)-alpha-[4-(1-methylethyl)phenyl]-1-piperidinebutanol, Fexofenadine Related Compound C, Fexofenadine Hydrochloride Impurity C, Fexofenadine Impurity C
InChI formel InChI=1S/C31H39NO2/c1-24(2)25-15-17-26(18-16-25)30(33)14-9-21-32-22-19-29(20-23-32)31(34,27-10-5-3-6-11-27)28-12-7-4-8-13-28/h3-8,10-13,15-18,24,29-30,33-34H,9,14,19-23H2,1-2H3
LEDER CC(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
IUPAC-namn 4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]-1-(4-propan-2-ylphenyl)butan-1-ol
Molekylvikt (g/mol) 457.65
Kvantitet 100 mg
Formel vikt 457.2981 g/mol