Beskrivning
FFAR2/GPR43 Polyclonal antibody specifically detects FFAR2/GPR43 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot
Specifikationer
Specifikationer
| Antigen | FFAR2/GPR43 |
| Användningsområden | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Utspädning | Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:150, Immunohistochemistry-Paraffin, Dot Blot 1:10000 |
| Formulering | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo- protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43FFA2R, G-protein coupled receptor 43 |
| Värd art | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 250-300 on human free fatty acid receptor 2 protein. |
| Reningsmetod | Affinity-purified |
| Visa mer |