Learn More
Sperian™ FFP1D Filtering Half Masks
425.52 SEK - 767.41 SEK
Specifications
Disposable Yes
Latex Free Yes
Material (Faceshield) PES knit, PU foam, PU film
Model FFP1D
Nose Piece Material Galvanized wire wrapped by PP
View More Specs
Products 2
Product Code Brand For Use With (Application) Price Quantity & Availability  
Product Code Brand For Use With (Application) Price Quantity & Availability  
11836960 View Documents Sperian™
1005593
Textile, Abattoirs, Metallurgy, Construction, Iron and Steel Industries, Hospitals, etc.. 767.41 SEK
Pack of 20 View alternative products
Save Up to 
»
Remote Stock
(10 days availability): 6
Add to basket
11891771 View Documents Sperian™
1005605
Textile, construction, iron and steel industries 425.52 SEK
Pack of 20 View alternative products
Save Up to 
»
Remote Stock
(10 days availability): 4
Add to basket
Specifications
Specifications
Yes
PES knit, PU foam, PU film
Galvanized wire wrapped by PP
Fluid
Filtering Half Mask for Single Use
Yes
FFP1D
20
Yes