Learn More
3M™ FFP2 Valved Particulate Respirator
3M™ FFP2 Valved Particulate Respirator provides FFP2 respiratory protection while adapting to the face shape for a comfortable fit.
Brand: 3M™ 7000062038
Additional Details : Weight : 2.78692kg
Description
- Provides FFP2 (NPF 12) protection against hazardous dusts and mists
- Classic cup-shaped respirator
- 3M™ Cool Flow™ Valve helps reduce heat build-up
- The lightweight, twin-strap design, nose foam and nose clip help provide a comfortable, custom fit
- Features a durable, collapse-resistant inner shell
- Designed to work well with 3M™ Eyewear and Hearing Protection products/li>
Specifications
CE Approved, EN 149: 2001 + A1: 2009 FFP2 NR D | |
White | |
Respirator | |
Cup |