missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17400-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.Specifications
FGF1 | |
Polyclonal | |
Unconjugated | |
FGF1 | |
AFGF, ECGF, ECGF beta, ECGFA, ECGFB, FGF alpha, FGF1, FGFA, GLIO703, HBGF 1, HBGF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14164, 2246, 25317 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05230, P61148, P61149 | |
FGF1 | |
FGF1 Fusion Protein Ag11366 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |