missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16876-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to FGF16.
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation, and is required for normal cardiomyocyte proliferation and heart development. .Specifications
FGF16 | |
Polyclonal | |
Unconjugated | |
FGF16 | |
FGF 16, FGF16, fibroblast growth factor 16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60464, 80903, 8823 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43320, O54769, Q9ESL8 | |
Fgf16 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |