missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550960
Additional Details : Weight : 0.01000kg
Description
FGF22 Polyclonal Antibody detects endogenous levels of FGF22 protein.
FGF22 Polyclonal antibody specifically detects FGF22 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
FGF22 | |
Polyclonal | |
Unconjugated | |
FGF22 | |
2210414E06Rik; FGF; Fgf22; FGF-22; Fibroblast growth factor; Fibroblast growth factor 22; UNQ2500/PRO5800 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170579, 27006, 67112 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9ESS2, Q9HCT0 | |
FGF22 | |
Synthesized peptide derived from the Internal region of human FGF-22. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |