missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24725-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
FGFBP3 is a heparin-binding protein which binds to FGF2, prevents binding of FGF2 to heparin and probably inhibits immobilization of FGF2 on extracellular matrix glycosaminoglycans, allowing its release and subsequent activation of FGFR signaling which leads to increased vascular permeability.Specifications
FGFBP3 | |
Polyclonal | |
Unconjugated | |
FGFBP3 | |
C10orf13, FGF binding protein 3, FGF BP3, FGFBP 3, FGFBP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
143282, 72514 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q1HCM0, Q8TAT2 | |
FGFBP3 | |
FGFBP3 Fusion Protein Ag20573 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |