missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FGFBP3 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823215
|
Proteintech
24725-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813215
|
Proteintech
24725-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FGFBP3 is a heparin-binding protein which binds to FGF2, prevents binding of FGF2 to heparin and probably inhibits immobilization of FGF2 on extracellular matrix glycosaminoglycans, allowing its release and subsequent activation of FGFR signaling which leads to increased vascular permeability.Specifications
FGFBP3 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q1HCM0, Q8TAT2 | |
143282, 72514 | |
FGFBP3 Fusion Protein Ag20573 | |
Primary | |
-20°C | |
FGFBP3 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C10orf13, FGF binding protein 3, FGF BP3, FGFBP 3, FGFBP3 | |
FGFBP3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |