missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGFR1, His Tag Recombinant Protein
18260.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_000595 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 50.4 kDa |
Gene Alias | FGFR1, H2, H3, H4 |
KinaseFamily | FGF Receptor Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15458413
|
Invitrogen™
PR4660A |
100μg |
18260.00 SEK
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174842
|
Invitrogen™
PV4105 |
1 mg |
79000.00 SEK
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The FGFR family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.
Specifications
NP_000595 | |
50.4 kDa | |
FGF Receptor Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.2mM EDTA, 0.05% Triton X-100, 5mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
FGFR1, H2, H3, H4 | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic |