missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGFR1 [V561M] Recombinant Human Protein

FGFR1 (also known as FLT2) is part of the Fibroblast Growth Factor Receptor family of four membrane-spanning tyrosine kinases (FGFR1-4). Recombinant human FGFR1 (399-822; V561M) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

Brand:  Invitrogen™ PV6345

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15418983

  • 64700.00 SEK / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FGFR1-4 serve as high-affinity receptors for 17 growth factors (FGF1-17). The FGF Receptor family plays an important role in multiple biological processes, including mesoderm induction and patterning, cell growth and migration, organ formation and bone growth. FGFR1 is alternatively spliced generating multiple splice variants that are differentially expressed during embryo development and in the adult body.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
74.0kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
aa 399-822
Recombinant
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol
1mg
FLT2, CEK, FLG, KAL2, BFGFR, C-FGR, CD331, N-SAM
Human
GST Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers