missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 [K650E] Recombinant Human Protein
FGFR3 [K650E] Recombinant Human Protein
Brand: Invitrogen PV4392
Additional Details : Weight : 0.00900kg
Description
FGFR3 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. The K650E mutation leads to constitutive activation and the neonatal lethal disorder TDII.Specifications
NP_000133; K650E | |
Cell Structure, Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
FGFR3 [K650E] Recombinant Human Protein | |
–80°C | |
1 Each | |
FGFR3 [K650E] Recombinant Human Protein | |
FGF receptor | |
FGFR3 [K650E] Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
0.5mg/mL | |
49.8kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
FGFR3 K650E | |
FGFR3 K650E was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2,1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 398-806; K650E | |
879nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag | |
Kinase and inhibitors | |
75% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.