Läs mer
Invitrogen™ Human FGFR3 [K650M], GST Tag, His Tag Recombinant Protein
Beskrivning
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Human FGFR3, C-terminal fragment, amino acids R397-T806 (as in NCBI/Protein entry NP_000133.1) with a K650M mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant
Specifikationer
Specifikationer
| Koncentration | See Label |
| För användning med (applikation) | Kinase Assay |
| Formulering | 50 mM HEPES with 100 mM NaCl, 15 mM glutathione, 20% glycerol, 5 mM DTT and no preservative; pH 7.5 |
| Molekylvikt (g/mol) | 73.9 kDa |
| Namn | Human FGFR3 [K650M], GST Tag, His Tag |
| Kvantitet | 10 μg |
| Vanligt namn | FGFR3 (K650M Mutant) |
| Protein Längd | 397-806 |
| Konjugera | Unconjugated |
| Art | Human |
| Visa mer |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.