missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Fibrinogen gamma Monoclonal Antibody (4H9)
GREENER_CHOICE

Produktkod. 12877922
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
12877922 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 12877922 Leverantör Invitrogen™ Leverantörsnummer MA515906

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

MA5-15906 targets FGG in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15906 immunogen is purified recombinant fragment of human FGG expressed in E. Coli. . MA5-15906 detects FGG which has a predicted molecular weight of approximately 52kDa.

The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Fibrinogen gamma
Användningsområden ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassificering Monoclonal
Klona 4H9
Koncentration Conc. Not Determined
Konjugera Unconjugated
Formulering Ascites with 0.03% sodium azide
Gen Fgg
Genaccessionsnr. P02679, Q8VCM7
Gene Alias 3010002H13Rik; AI256424; FGG; fibg; fibrinogen; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma'; gamma-fibrinogen; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951)
Gensymboler Fgg
Värd art Mouse
Immunogen Purified recombinant fragment of human FGG expressed in E. Coli.
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 2266, 99571
Målarter Human, Mouse
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG2a
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.