missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Fibulin 2 Monoclonal Antibody (7H4F6/3B9A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515390
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA5-15390 targets FBLN2 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15390 immunogen is purified recombinant fragment of FBLN2 (aa180-440) expressed in E. Coli.
This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifikationer
| Fibulin 2 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P98095 | |
| FBLN2 | |
| Purified recombinant fragment of FBLN2 (aa180-440) expressed in E. Coli. | |
| RUO | |
| 2199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 7H4F6/3B9A11 | |
| Unconjugated | |
| FBLN2 | |
| 5730577E14Rik; FBLN2; FIBL2; FIBL-2; fibulin 2; Fibulin2; fibulin-2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering