missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficoll™, MP Biomedicals™
Click to view available options
Quantity:
5 g
25 g
Specifications
Color | White |
Physical Form | Powder |
Density | <1.2 g/mL |
Quantity | 5 g |
Formula Weight | ≈400,000 |
Chemical Name or Material | Ficoll™ |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'