missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FICOLL™, White Powder, MP Biomedicals™
Click to view available options
Quantity:
10 g
100 g
Specifications
Color | White |
Density | up to 1.2g/mL |
Quantity | 10 g |
Formula Weight | ≈400,000 |
Physical Form | Solid |
Chemical Name or Material | FICOLL™ |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'