Läs mer
Tocris Bioscience™ Filipin III
Beskrivning
Filipin III is a polyene antibiotic. Binds sterols in fungal membranes. Also inhibits formation of the pathological form of prion protein (PrP-res) from the normal membrane-bound (PrP-sen) form. Also fluorescent cholesterol stain.
Specifikationer
Specifikationer
| CAS | 480-49-9 |
| Kvantitet | 1 mg |
| Molekylformel | C35H58O11 |
| Formel vikt | 654.83g/mol |
| Produkttyp | Inhibitor |
For Research Use Only