missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Candle with Glass Tube, Conical Shape

Filter Candle with narrow glass tube.

Brand:  ROBU VitraPOR™ 18301

Additional Details : Weight : 0.13020kg

 View more versions of this product

Product Code. 16812966

  • 1365.00 SEK / Pack of 2
Estimated Shipment: 03-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
Glass Tube, Ø 13 mm
65 mm
1 μm
1
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers