missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Filter Candle with Glass Tube, Conical Shape
Filter Candle with narrow glass tube.
Brand: ROBU VitraPOR™ 18313
Additional Details : Weight : 0.15680kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass 3.3 | |
3 | |
25 mm |
Glass Tube, Ø 20 mm | |
65 mm |