missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Filter Cellulose Nitrate
Brand: Cytiva 10405076
Additional Details : Weight : 0.01000kg
Specifications
47 mm | |
Cellulose Nitrate |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- AE99/5 membrane, cellulose nitrate, 8µm 47mm HYRIM
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose