missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Filter Crucible, 15 mL
Crucibles are marked with an individual number and allow a distinct sample.
1540.00 SEK - 3295.00 SEK
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
16872996
|
ROBU VitraPOR™
20151 |
1 |
1540.00 SEK
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16882996
|
ROBU VitraPOR™
20152 |
2 |
1540.00 SEK
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16813006
|
ROBU VitraPOR™
20155 |
5 |
3295.00 SEK
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16803006
|
ROBU VitraPOR™
20154 |
4 |
1790.00 SEK
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16862996
|
ROBU VitraPOR™
20150 |
0 |
1540.00 SEK
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16892996
|
ROBU VitraPOR™
20153 |
3 |
1540.00 SEK
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
- For analytical filtration in laboratory
- Suitable glass adapters and rubber sleeves are available
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
15 mL |