missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Crucible, 8 mL

Crucibles are marked with an individual number and allow a distinct sample.

Brand:  ROBU VitraPOR™ 20082

Additional Details : Weight : 0.17950kg

 View more versions of this product

Product Code. 16822996

  • 1203.60 SEK / Pack of 10

Estimated Shipment: 10-12-2021
Add to basket

Description

Description

  • For analytical filtration in laboratory
  • Suitable glass adapters and rubber sleeves are available
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

8 mL
Borosilicate Glass 3.3
2
Videos
SDS
Documents
Certificates
Special Offers

Special Offers