Learn More
ROBU VitraPOR™ Filter Crucible, CFE
Filter Crucible with individual number to allow distinct sample identification.
Brand:  ROBU VitraPOR™ 20600
Additional Details : Weight : 0.37810kg
 View more versions of this product
Product Code. 16883016
  • 2295.00 SEK / Pack of 10
Estimated Shipment: 17-04-2023
to see stock.
Add to basket
Description
Description
  • For crude fiber extraction for many current systems
  • The VitraPOR™ filter discs have an improved sintering hardness to withstand the chemical load in the analytical processes
Specifications
Specifications
Borosilicate Glass 3.3
10/Pk.
30 mL
0