Learn More
ROBU VitraPOR™ Filter Crucible, CFE
Filter Crucible with individual number to allow distinct sample identification.
Brand: ROBU VitraPOR™ 20600
Additional Details : Weight : 0.37810kg
Description
- For crude fiber extraction for many current systems
- The VitraPOR™ filter discs have an improved sintering hardness to withstand the chemical load in the analytical processes
Specifications
Borosilicate Glass 3.3 | |
10/Pk. | |
30 mL | |
0 |