Learn More
BRAND™ Filter Crucibles
Borosilicate glass construction. Squat form offers stability and low risk of spillage.
Brand:  BRAND™ 458733
4469.84 SEK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 4.96000kg
 View more versions of this product
Product Code. 15252449
  • 5005.00 SEK / Pack of 10
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
  • Borosilicate glass construction
  • Supplied in packs of 10
Specifications
Specifications
Borosilicate Glass
50 mL
Filter crucible
3