Learn More
ROBU VitraPOR™ Filter Disc with 8.5 mm Bore, Ground Cylindrical
Filter Disc for use with gas distribution tubes and washing bottle heads.
Brand:  ROBU VitraPOR™ 17000
Additional Details : Weight : 0.05040kg
 View more versions of this product
Product Code. 16882886
  • 476.34 SEK / Pack of 10
Estimated Shipment: 14-12-2021
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass 3.3
23 mm
0
Non-sterile
3.0 ± 0.5 mm
Bore, Ø 8.5 mm