missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 150 mm

The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.

Brand:  ROBU VitraPOR™ 61501

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 16823216

  • 1943.10 SEK / Each

Estimated Shipment: 17-12-2021
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications

Specifications

1 μm
Non-sterile
8.5 ± 0.5 mm
1 Ea.
Borosilicate Glass 3.3
150 mm
1
Videos
SDS
Documents
Certificates
Special Offers

Special Offers