missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 160 mm

The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.

Brand:  ROBU VitraPOR™ 61601

Additional Details : Weight : 0.38000kg

 View more versions of this product

Product Code. 16883216

  • 2735.00 SEK / Each
Estimated Shipment: 15-04-2024
to see stock.
Add to basket

Description

Description

IFNA1 Monoclonal Antibody for ELISA

  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications

Specifications

Borosilicate Glass (3.3)
Non-sterile
1
160 mm
1 μm
1 Ea.
9.5 ± 0.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers