Learn More
ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 20 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Brand:  ROBU VitraPOR™ 16209
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16822826
  • 632.40 SEK / Pack of 10
Estimated Shipment: 20-12-2021
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications
Specifications
00
Non-sterile
2.5 ± 0.2 mm
10/Pk.
Borosilicate Glass 3.3
20 mm
00