missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 290 mm

The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.

Brand:  ROBU VitraPOR™ 62903

Additional Details : Weight : 2.13750kg

 View more versions of this product

Product Code. 16843286

  • 13700.00 SEK / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications

Specifications

Borosilicate Glass (3.3)
3
290 mm
Non-sterile
18.0 mm ± 0.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers