missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 80 mm

The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.

Brand:  ROBU VitraPOR™ 16800

Additional Details : Weight : 0.24220kg

 View more versions of this product

Product Code. 16822876

  • 1940.00 SEK / Pack of 5

Estimated Shipment: 10-12-2021
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications

Specifications

Borosilicate Glass 3.3
80 mm
0
Non-sterile
5.0 ± 0.2 mm
5/Pk.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers