Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 10 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand: ROBU VitraPOR™ 15104
Additional Details : Weight : 0.01000kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
10/Pk. | |
2.8 ± 0.7 mm |
Non-sterile | |
4 | |
10 mm |